This item is now discontinued. Please browse similar items

Ibi Scientific Histochemistry Staining Tray, 12-place, clear lid

Representative image only.
Ibi Scientific Mfr # ATC-12 – Item # RK-00485-92
Designed specifically for immunohistochemistry slide staining

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Number of slides)12
  • External Width (in)7.5
  • External Height (in)1
  • External Depth (in)10 1/2
  • External Width (cm)19.05
  • External Height (cm)2.54
  • External Depth (cm)26.67
  • DescriptionHistochemistry Staining Tray, 12-place, clear lid

More About this Item

These staining trays work well in microscopy, cytology, and hematology settings and are designed for immunohistochemistry assay slide staining in the lab. All trays have an integrated dual-well design reservoir to hold water beneath the slides when humidity is needed. The skid-proof rubber feet prevent sliding and allow for stable stacking for a tidy benchtop.

Trays have a clear lid for easy visibility of the slides.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat