This item is now discontinued. Please browse similar items
Honeywell NXLO404-MED-090 NextGen Athletic Alloy Toe Shoes, Size 9; 1 Pair
Representative image only.
Honeywell Mfr # NXLO404-MED-090 – Item # RK-86326-97
The next generation of work shoe performance
  • Heat-resistant outsole with excellent slip resistance
  • Durable—waterproof leather construction
  • No exposed hardware for your protection
  • Meet ASTM F2413-11 M I/75 C/75 EH requirements
  • Available in men sizes 7 to 13, medium width
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Size9
  • ColorGray
  • Men's Size9
  • ComplianceASTM F2413-11 M I/75 C/75 EH
  • DescriptionNextGen Athletic Alloy Toe Shoes, Size 9; 1 Pair
Key Features
  • Heat-resistant outsole with excellent slip resistance
  • Durable—waterproof leather construction
  • No exposed hardware for your protection
  • Meet ASTM F2413-11 M I/75 C/75 EH requirements
  • Available in men sizes 7 to 13, medium width
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
NextGen work shoes feature a unique blend of waterproof leather, polyurethane (PU) coated leather, and ballistic nylon, giving you the performance you need. The outsole is heat-resistant up to 356°F (180°C), while providing SRC-rated slip resistance. Multiple layers of multi-density polyurethane foam provide airflow, comfort, and ergonomic support. No exposed hardware and an alloy toe offer all-day protection.