This item is now discontinued. Please browse similar items
Honeywell Durafet III Non-Glass pH Electrode, 3/4" In-Line Threads
Representative image only.
Honeywell Mfr # 07777DVP-03-02-0000-000 – Item # RK-56611-22
Provides accurate and stable pH measurements
  • Rugged design reduces replacement
  • Faster than glass electrodes
  • Ideal for in-line applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRyton PPS
  • Compatible WithHoneywell Multiple-Input Controller
  • Connections3/4\" NPT(M)
  • Length (in)600
  • DescriptionDurafet III, non-glass pH electrode, 3/4 inch in-line threads
  • Warranty1 year
1 Year Warranty
Key Features
  • Rugged design reduces replacement
  • Faster than glass electrodes
  • Ideal for in-line applications
  • 1 year warranty
More About this Item
Durafet III non-glass pH electrode features a durable Ryton PPS body with a 3/4" NPT(M) process connection, and a quick disconnect 50-ft (15-m) cable. The pH measurement is based on ISFET (Ion Selective Field Effect Transistor) technology, which produces an electrode up to 10 times faster than glass electrodes. Fast response improves product quality and provides better process control to optimize chemical usage.