This item is now discontinued. Please browse similar items
Honeywell Coreshield™ 13 Gauge ANSI A3/C Glove, 7/S
Representative image only.
Honeywell Mfr # 23-0513B/7S – Item # RK-86326-60
Sturdy cut-resistant gloves simplify selection for a variety of applications
  • Color-coded marking system highlights the right gloves for your application, per EN/ANSI standards
  • Choose by level of cut resistance, protection, grip, dexterity and fit
  • One of the lightest-weight cut-protective gloves on the market
  • Touchscreen resistive and capacitive
  • Water and oil proof coating
  • Size 7/S
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size7/S
  • ColorBlack/Green
  • LiningHPPE/basalt
  • ApprovalANSI A3, ISO level C
  • DescriptionHoneywell Coreshield™ 13 Gauge ANSI A3/C Glove, 7/S
Key Features
  • Color-coded marking system highlights the right gloves for your application, per EN/ANSI standards
  • Choose by level of cut resistance, protection, grip, dexterity and fit
  • One of the lightest-weight cut-protective gloves on the market
  • Touchscreen resistive and capacitive
  • Water and oil proof coating
  • Size 7/S
More About this Item
Providing durability and dexterity, value-packed CoreShield™ gloves from Honeywell offer optimum cut protection and user comfort. The lightweight gloves prevent hand fatigue, while a specially designed microfoam elastic coating provides outstanding oil- and dry-grip performance.
Each glove is made with softer, skin-friendly yarns that--compared to glass fiber--reduce the allergy risk to wearers. Due to this, all CoreShield™ gloves are certified STANDARD 100 by OEKO-TEX®.
CoreShield™ coatings offer 360° breathability through an open cell structure, further enhancing comfort even upon extensive wear. The washability of the glove material means little change in performance or size after five washes.