This item is now discontinued. Please browse similar items
Honeywell CB03 Circuit Breaker lockouts; single-pole breakers, 6/pack
Representative image only.
Honeywell Mfr # CB03 – Item # RK-86200-30
SPECS & DESCRIPTION
Specifications & Description
  • Locks OutSingle-pole \"no hole\" breakers (115 or 277 volts)
  • DescriptionCircuit Breaker lockouts; single-pole breakers, 6/pack
More About this Item
Lock breakers in “off” position without electrical or mechanical alterations. Circuit breaker lockouts fit most major brand single-pole (86200-30) or double-pole (86200-32) breakers. No self-locking tie required. Pack of six padlocks can be ordered separately.
C-Safe—TM North Safety Products