This item is now discontinued. Please browse similar items
Honeywell B100200 Respirator Cartridges, Acid Gases, 6/box
Representative image only.
Honeywell Mfr # B100200 – Item # RK-81662-72
Screw-on cartridges install easily on respirators
SPECS & DESCRIPTION
Specifications & Description
  • RatingNone
  • NIOSH ApprovalChlorine, Hydrogen Chloride, Sulfur Dioxide, Formaldehyde
  • ComplianceNIOSH
  • ColorWhite
  • DescriptionRespirator Cartridges, Acid Gases, 6/box
More About this Item
NIOSH approved for use with chlorine, hydrogen chloride, sulfur dioxide, and formaldehyde.