This item is now discontinued. Please browse similar items
Honeywell 322500 Half-Mask Respirator, Medium/Large
Representative image only.
Honeywell Mfr # 322500 – Item # RK-81662-64
Silicone skirt is flexible, lightweight, and durable
  • Triple-flange design provides the ultimate in protection and comfort
SPECS & DESCRIPTION
Specifications & Description
  • SizeMedium/large
  • ComplianceNIOSH
  • ColorBlack
  • DescriptionHalf-Mask Respirator, Medium/Large
Key Features
  • Triple-flange design provides the ultimate in protection and comfort
More About this Item
These lightweight respirators offer a comfortable, secure fit for excellent protection against organic vapors, acid gases, contaminates, and more. Choose from several styles of cartridges and filters (order separately below) according to the demands of your work environment.
Approval: NIOSH
Survivair Premier® Plus Half-Mask Respirators feature a wide, wraparound facepiece that comfortably fits a variety of facial types. Lateral positioning of cartridges enhances field of vision.