This item is now discontinued. Please browse similar items
Honeywell 31-70102 Laser Goggles, Vented, Uvextreme Coated; YAG/KTP
Representative image only.
Honeywell Mfr # 31-70102 – Item # RK-86528-03
Superior side protection
  • Translucent goggle body offers same protection as the lens
  • Soft comfortable polymer conforms to individual facial contours for proper fit
  • Vent system for air flow
  • Uvextreme® coating reduces fogging, scratches, and static
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorWhite
  • Lens ColorBrown
  • Lens CoatingUvextreme® (reduces fogging, scratches, and static)
  • Laser TypeYAG, KTP
  • ComplianceANSI Z136.1-1986, Z87.1-2003
  • DescriptionLaser Goggles, Vented, Uvextreme Coated; YAG/KTP
Key Features
  • Translucent goggle body offers same protection as the lens
  • Soft comfortable polymer conforms to individual facial contours for proper fit
  • Vent system for air flow
  • Uvextreme® coating reduces fogging, scratches, and static
More About this Item
Compliance: ANSI Z136.1-1986, Z87.1-2003