This item is now discontinued. Please browse similar items
Honeywell 31-40135 Laser Safety Glasses, HeNe/Red Diode
Representative image only.
Honeywell Mfr # 31-40135 – Item # RK-86438-28
Excellent visible light transmission minimizes distortion
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorBlue
  • Laser TypeHelium Neon (HeNe), red diodes
  • ComplianceANSI Z136.1-1986, Z87.1-2003
  • WavelengthOD 1-2 at 630-680 nm
  • DescriptionLaser Safety Glasses, HeNe/Red Diode
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
LOTG Series
Eyewear features a broad, single-lens design to ensure maximum top and side protection. Glasses even fit comfortably over most prescription glasses.
Compliance: ANSI Z136.1-1986, Z87.1-2003 (glasses unsuitable for direct laser radiation).