This item is now discontinued. Please browse similar items
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 11; 1 Pair
Representative image only.
Honeywell Mfr # 22114-CTM-110 – Item # RK-86326-85
Keep workers’ feet dry and comfortable
  • Triple dipped in neoprene for 100% waterproof boots
  • Chemical-resistant neoprene construction
  • Electrical hazard rated outsole and steel toe impact protection
  • Meet ASTM F2413-18 M/I/C EH requirements
  • Available in mens' sizes 4 to 13
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)12
  • MaterialNeoprene
  • ColorCopper & tan
  • Men's Size11
  • ComplianceASTM F2413-18 M/I/C EH
  • DescriptionNeoprene Steel Toe Men's Work Boots, Size 11
Key Features
  • Triple dipped in neoprene for 100% waterproof boots
  • Chemical-resistant neoprene construction
  • Electrical hazard rated outsole and steel toe impact protection
  • Meet ASTM F2413-18 M/I/C EH requirements
  • Available in mens' sizes 4 to 13
More About this Item
Servus® by Honeywell™ work boots feature hand-laid natural rubber to offer flexibility and strength at critical stress points. The boots are then triple dipped in neoprene and vulcanized to create a 100% seamless, liquid-tight boot. Self-cleaning chevron outer sole has a flat, surface-to-surface tread pattern for increased slip resistance on slick surfaces and the open channels keep out liquids and debris. The work boots meet ASTM F2413-18 hazard and steel toe (M 1/75 & C75) requirements. The removable and washable Breath-O-Prene® liner absorbs shocks and keeps away moisture to keep your feet dry and comfortable.