This item is now discontinued. Please browse similar items
Honeywell 14110444 N95 Particulate Respirators, 20/box
Representative image only.
Honeywell Mfr # 14110444 – Item # RK-81660-40
Molded cup contours with the natural shape of the face, minimizing pressure points
  • Dual headstraps and soft inner lining further increase comfort for long term use
SPECS & DESCRIPTION
Specifications & Description
  • RatingN95
  • ComplianceNIOSH
  • ColorWhite
  • DescriptionN95 Particulate Respirators, 20/box
Key Features
  • Dual headstraps and soft inner lining further increase comfort for long term use
More About this Item
Universal one-size respirators are designed to fit most face shapes and sizes. Durable outer shell prevents the mask from collapse when working in high-humidity environments. Molded-nose bridge ensures a comfortable fit and works well with glasses. Electrostatic polypropylene/polyester fibers provide excellent filter performance in applications such as sanding wood, dust exposure, grinding, metal cutting, and healthcare.
Approval: NIOSH