This item is now discontinued. Please browse similar items
Honeywell 14110430 Particulate Respirators, P95/Organic Vapor/Valve/M-L; 10/Bx
Representative image only.
Honeywell Mfr # 14110430 – Item # RK-81660-25
Three sizes for maximum protection, color-coded nose seal for easy size recognition
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
SPECS & DESCRIPTION
Specifications & Description
  • RatingN95
  • SizeMedium/large
  • ComplianceNIOSH
  • DescriptionParticulate Respirators, P95/Organic Vapor/Valve/M-L; 10/Bx
Key Features
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
More About this Item
Use these respirators when working in areas containing oil-free particulate aerosols. These compact respirators are easy to use.
Approval: NIOSH
Models with exhalation valves ensure comfortable breathing even in heated environments. Organic vapor and acid gas respirators protect you from nuisance-level gases that do not exceed OSHA permissible exposure limit.