This item is now discontinued. Please browse similar items
Honeywell 14110390 Particulate Respirators; N95, Standard, Small, Box of 20
Representative image only.
Honeywell Mfr # 14110390 – Item # RK-81660-00
Three sizes for maximum protection, color-coded nose seal for easy size recognition
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
SPECS & DESCRIPTION
Specifications & Description
  • RatingN95
  • SizeSmall
  • ComplianceNIOSH
  • ColorWhite
  • DescriptionParticulate Respirators; N95, Standard, Small, Box of 20
Key Features
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
More About this Item
Use these respirators when working in areas containing oil-free particulate aerosols. These compact respirators are easy to use.
Approval: NIOSH
Standard models are ideal for grinding, sanding, sweeping, and other work in dusty environments.