This item is now discontinued. Please browse similar items
Honeywell 14110094CC Nuisance Dust Mask, Box of 50
Representative image only.
Honeywell Mfr # 14110094CC – Item # RK-81660-80
  • Economical particulate protection
SPECS & DESCRIPTION
Specifications & Description
  • ColorWhite
  • DescriptionNuisance Dust Mask, Box of 50
Key Features
  • Economical particulate protection
More About this Item
Limit exposure to food particles, construction dusts, pollen, or any other nuisance particulates that are present at levels below the OSHA Permissible Exposure Limit (PEL). Not NIOSH-approved.