This item is now discontinued. Please browse similar items
Honeywell 105810 Respirator Cartridges, Multi-Contaminate, with P100 filter, 4/box
Representative image only.
Honeywell Mfr # 105810 – Item # RK-81662-77
Screw-on cartridges install easily on respirators
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Ammonia/Methylamine, Hydrogen Sulfide (escape only), Formaldehyde, Nitrogen Dioxide
  • ComplianceNIOSH
  • ColorGreen, Pink
  • DescriptionRespirator Cartridges, Multi-Contaminate, with P100 filter, 4/box
More About this Item
NIOSH approved for use with organic vapors, ammonia, methylamine, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen fluoride, hydrogen sulfide (esc), formaldehyde, and nitrogen dioxide. Includes a P100 prefilter.