This item is now discontinued. Please browse similar items
Honeywell 105210 Respirator Cartridges, Acid Gases, with P100 filter, 4/box
Representative image only.
Honeywell Mfr # 105210. – Item # RK-81662-73
Screw-on cartridges install easily on respirators
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalChlorine, Hydrogen Chloride, Sulfur Dioxide, Formaldehyde
  • ComplianceNIOSH
  • ColorPink, White
  • DescriptionRespirator Cartridges, Acid Gases, with P100 filter, 4/box
More About this Item
NIOSH approved for use with chlorine, hydrogen chloride, sulfur dioxide, and formaldehyde. Includes a P100 prefilter.