This item is now discontinued. Please browse similar items
Elcometer Highline Meter II Highline Meter II Paint and Thickness Coating Gauge; 0-40 mils
Representative image only.
Elcometer Mfr # Highline Meter II – Item # RK-54102-02
Meet industry standards and regulations ensuring compliance
  • Reversible display allows you to read measurements when gauge is inverted
  • Lightweight rugged design is ideal for the lab or manufacturing floor
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 40 mil
  • Resolution0.1 mil (1 µm)
  • Accuracy± (3% + 0.1 mil or 2 µm)
  • ScaleFerrous/Nonferrous
  • DisplayLCD
  • PowerOne AAA battery
  • Depth (in)1
  • DescriptionPaint and Thickness Coating Gauge; 0-80 mils
  • Warranty1 year
1 Year Warranty
Key Features
  • Reversible display allows you to read measurements when gauge is inverted
  • Lightweight rugged design is ideal for the lab or manufacturing floor
  • 1 year warranty
More About this Item
This paint and coating thickness gauge measures thickness of coatings on ferrous and non-ferrous metals. Accurate and economical, this gauge is an ideal instrument for paint applicators, coating inspectors and more. Unit is designed with an embedded v-groove probe for easy positioning on cylindrical parts and surfaces and features audible measurement indication notifying operator of measurement completion.
The oversized LCD display is reversible making it simple to read measurements when the gauge is inverted. Gauge includes a zero feature for rough or curved surfaces and a handy reset feature for use when no zero reference is available. This gauge does not require calibration for most applications and features mils/micron user selectable capability for additional application flexibility.