High-Molecular Weight HDPE Bags, Clear, 30 gal; 500/Pk
Representative image only.
Item # RK-60104-04
$150.65CAD / Pkg of 500
BUY MORE & SAVE
  • 1-5$150.65CAD / Pkg of 500
  • 6-29$135.59CAD / Pkg of 500
  • 30-119$128.05CAD / Pkg of 500
  • 120+$120.52CAD / Pkg of 500
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Gal)30
  • MaterialHDPE
  • ColorClear
  • Length (in)30
  • Width (in)37
  • Length (cm)76.2
  • Width (cm)93.98
  • Thickness (mil)0.39
  • DescriptionHigh-Molecular Weight HDPE Bags, Clear, 30 gal; 500/Pk
More About this Item
These high-molecular weight HDPE bags have excellent impact and tensile strength. Is FDA compliant for use in food contact applications. Bags 60104-00 through -16 are clear; bag 60104-18 is metallic silver.