This item is now discontinued. Please browse similar items
High Five Industrial Nitrile Gloves, Small, 100/box
Representative image only.
High Five Mfr # N251 – Item # RK-86233-15
Durable glove is thicker for tougher tasks
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5.5
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • GripTextured
  • CuffBeaded
  • SterileNo
  • DescriptionIndustrial Nitrile Gloves, Small, 100/box
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Powder-free nitrile gloves are ideal for industrial and food processing applications. Glove surface is textured for a more secure grip; beaded cuff. Disposable; for single-use only. Ambidextrous.