This item is now discontinued. Please browse similar items
HF Scientific 52520 Hach ProCal Primary Standard for Portable Turbidimeter 2100Q, 0.0 NTU
Representative image only.
HF Scientific Mfr # 52520 – Item # RK-99900-48
Instrument specific kits and standards ensure maximum accuracy
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • NTU Value0.0
  • Volume (mL)500
  • Volume (oz)16.9
  • Use withHach 2100P Portable Turbidimeters
  • DescriptionHach ProCal Primary Standard for Portable Turbidimeter 2100Q, 0.0 NTU
  • Warranty1 year
1 Year Warranty
Key Features
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
More About this Item
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
One 500-mL bottle