This item is now discontinued. Please browse similar items
HF Scientific 52040 Hach ProCal Primary Standard Kit for Process Turbidimeter 1720
Representative image only.
HF Scientific Mfr # 52040 – Item # RK-99900-33
Instrument specific kits and standards ensure maximum accuracy
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • NTU Value0.0, 20
  • Volume (mL)1000
  • Volume (oz)33.8
  • Use withHach 1720 Process Turbidimeters
  • DescriptionHach ProCal Primary Standard Kit for Process Turbidimeter 1720
  • Warranty1 year
1 Year Warranty
Key Features
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
More About this Item
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
Four 1-L bottles of each (8 total)