This item is now discontinued. Please browse similar items
HD Electric DVM-80 Dual Stick Digital Voltmeter, True RMS; 5V to 40kV
Representative image only.
HD Electric Mfr # DVM-80 – Item # RK-23022-48
Get safe and accurate voltage readings
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)5
  • DC Max (Volts)40000
  • AC Min (Volts)5
  • AC Max (Volts)40
  • DescriptionDual Stick Digital Voltmeter, True RMS; 5V to 40kV
  • Warranty1 Year
1 Year Warranty
Key Features
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
  • 1 year warranty
More About this Item
The DigiVolt® series of digital voltmeters/phasing sets are designed to take voltage measurements and determine if two conductors are in-phase or out-of-phase on system voltages.
The DVM-80† is a dual-stick, dual-ranging voltmeter/phaser measuring voltages from 5 V to 40 kV. The two-stick design can be used in both overhead and underground applications and incorporates Phase Accurate Technology—making all voltage readings, whether made from line-to-line or line-to-ground, accurate and repeatable to within 1%.
The DVM-80UVM is a compact, high accuracy dual-stick voltmeter and phasing set for virtually all overhead and underground applications up to 40 kV. This is the same as the DVM-80, but with the added features of capacitive test point and peak hold modes.
†Dual-stick meters and kit will require two hotsticks; one for each probe.