Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 25 µL
Representative image only.
Hamilton Mfr # 80401 – Item # RK-07938-21
Achieve precise liquid delivery
  • Constructed of borosilicate glass
  • Syringe features a 2000 psi max pressure
  • 18 month warranty
PRODUCT OPTIONS
$94.30CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Tip
  • Syringe Capacity (µL)25
  • Needle IncludedNo
  • Needle GaugeNeedle not included
  • MaterialSST, borosilicate glass
  • Outside Diameter (OD) (mm)7.75
  • DescriptionMicroliter Low-Volume Syringe, Luer Tip, No Needle, 25 µL
  • Warranty18 months
18 Month Warranty
Key Features
  • Constructed of borosilicate glass
  • Syringe features a 2000 psi max pressure
  • 18 month warranty
More About this Item
Hamilton® 700-series microliter syringes are designed to solve the general liquid handling requirements of manufacturing and research laboratories. Syringes are used for manual and automated GC and HPLC injections and for everyday applications that require accurate liquid measuring and dispending.
Choose from cemented needle, removable needle, or Luer tip syringes. Syringes with luer tip connections do not include a needle, order needles separately.