Hamilton Gastight® Syringe with Luer Tip, 25 µL, 1000 psig
Representative image only.
Hamilton Mfr # 80201 – Item # RK-07939-21
Gas- and liquid-tight up to 200 psi!
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Sample loss is negligible at pressures up to 200 psi
  • Gastight® syringe features a 1000 psig max pressure
  • 18 month warranty
PRODUCT OPTIONS
$98.86CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Tip
  • Syringe Capacity (mL)0.025
  • Syringe Capacity (µL)25
  • Needle IncludedNo
  • MaterialBorosilicate glass, PTFE
  • Outside Diameter (OD) (mm)7.75
  • DescriptionSyringe with Luer Tip, 25 µL, 1000 psig
  • Warranty18 months
18 Month Warranty
Key Features
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Sample loss is negligible at pressures up to 200 psi
  • Gastight® syringe features a 1000 psig max pressure
  • 18 month warranty
More About this Item
These essentially inert syringes have barrels and plungers that are interchangeable within each syringe size, allowing field repair. Syringes with Luer Tip Connections feature a ground glass, luer taper. Autoclavable if plunger is removed. Needles are not included, order separately.
Not for use in the withdrawal mode when using syringe pumps.