This item is now discontinued. Please browse similar items
Hach 3422E3A Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=10.0
Representative image only.
Hach Mfr # 3422E3A – Item # RK-19065-68
Contacting conductivity measurements from ultra-pure to high conductivity measurements
  • Suitable for GLI Contacting Conductivity Controller and Transmitter
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 10
  • Cell TypeContact
  • Electrode MaterialTitanium
  • Body MaterialPVDF
  • Fitting¾\" NPT(M)
  • Mounting TypeThreaded
  • Min temperature (° C)-5
  • Max temperature (° C)150
  • Min temperature (° F)23
  • Max temperature (° F)302
  • Cable Length (m)6
  • Cable Length (ft)20
  • DescriptionContacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=10.0
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for GLI Contacting Conductivity Controller and Transmitter
  • 1 year warranty
More About this Item
Nonmetallic Ryton® PPS cells 19065-55 and -57 have graphite electrode with 3⁄4" NPT(M) threads for mounting onto a 3⁄4" pipe tee or immersion applications. Sensor includes built-in 1000 Ω Pt RTD and 20-ft (6-m) cable.
PVDF cells 19065-60 through -68 have titanium electrodes with either 1⁄2" or 3⁄4" NPT(M) compression fitting for mounting onto a pipe tee with up to four inches of insertion depth. Reversing the compression fitting enables the sensor to be fastened onto the end of an immersion pipe. Cells include built-in 1000 Ω Pt RTD and 20-ft (6-m) cable.