Hach 225100 Color Disc Test Kit , Sulfate; 100 Tests/Kit
Representative image only.
Hach Mfr # 225100 – Item # RK-05545-95
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$213.64CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestSulfate
  • Measurement Points50 to 200 ppm
  • Tests Per Kit100
  • DescriptionColor Disc Test Kit , Sulfate; 100 Tests/Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.