Great Plains Industries 113255-4 Water Flowmeter/Totalizer, 3-30 GPM, 1" NPT(F), Nylon
Representative image only.
Great Plains Industries Mfr # 113255-4 – Item # RK-05611-22
Easy-to-read LCD displays batch or cumulative totals
  • Simple, small, and sturdy
  • Mount on hose or in-line
PRODUCT OPTIONS
$331.51CAD / Each
$456.75CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±5% of reading
  • Accuracy ±5.0
  • Max Operating Temperature (° F)131
  • Max Operating Temperature (° C)55
  • Fluid Typewater and thin fluids
  • BatteryTwo AA batteries
  • Battery Life5000 hours
  • Display Type6-digit LCD, 0.625"H
  • DescriptionWater Flowmeter/Totalizer, 3-30 GPM, 1" NPT(F), Nylon
Key Features
  • Simple, small, and sturdy
  • Mount on hose or in-line
More About this Item
Measure batch and cumulative totals in liquid transfer systems with these sturdy meters. Nylon meters are designed to measure water. Aluminum meters are designed to measure petroleum fuels. Meters can be used on any pump, pressure, or gravity feed systems. The batch total can be reset to measure flow for a single use. The cumulative total will automatically be reset to zero when the maximum reading of 9999 is met. Water models are nylon; fuel models are aluminum.