This item is now discontinued. Please browse similar items
Graphic Controls 10498484 Circular Chart, American Meter/Barton, 24 hour
Representative image only.
Graphic Controls Mfr # 10498484 – Item # RK-49000-54
Get exacting performance that meets American Gas Association (AGA) standards
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 15,000
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed1 day
  • Chart Diameter (in)12
  • DescriptionCircular Chart, American Meter/Barton, 24 hour
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 15,000
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.