This item is now discontinued. Please browse similar items
Graphic Controls 01166719 Circular Chart, 7 day
Representative image only.
Graphic Controls Mfr # 01166719 – Item # RK-49000-48
Get exacting performance that meets American Gas Association (AGA) standards
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scales read 0 to 100 and 0 to 500
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)12
  • Pen ColorBlue
  • DescriptionCircular Chart, 7 day
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scales read 0 to 100 and 0 to 500
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.