DWK Life Sciences (Kimble) Glass Babcock Bottles, Ice Cream Test, 18 gram Sample; 12/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # 1025-10 – Item # RK-34590-10
$654.19CAD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • AutoclavableYes
  • Overall Height (in)6 1/2
  • DescriptionGlass Babcock Bottles, Ice Cream Test, 18 gram Sample; 12/cs
More About this Item
Easily measure the percentage of butter fat content in your sample with these specialized bottles. The body of each bottle is a reaction chamber, while the graduated neck is a measuring device. Use the Babcock bottles for measuring liquids and small samples. The Paley bottles feature a large opening on the side of the body to easily accommodate larger or viscous samples. Replacement stoppers are available: please see accessories. All bottles measure 6 1/2" H.