This item is now discontinued. Please browse similar items
Glas-Col 100D SI02 Stirring/Heating Mantle Silicone Spillproof Guard; 2L
Representative image only.
Glas-Col Mfr # 100D SI02 – Item # RK-36226-93
Prevents damage from chemical spillage
  • Improves operating safety when heating liquids
  • Rugged silicone pad holds guard securely in place
  • Accomodates 2L spherical flasks
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Flask Size (mL)2000
  • DescriptionStirring/Heating Mantle Silicone Spillproof Guard; 2L
  • Warranty1 Year
1 Year Warranty
Key Features
  • Improves operating safety when heating liquids
  • Rugged silicone pad holds guard securely in place
  • Accomodates 2L spherical flasks
  • 1 year warranty
More About this Item
These spillproof guards prevent mantle damage from chemical spillage and improve operating safety when heating liquids. The guard is appropriate for use with low-wattage mantles for flask. A rugged silicone pad grips outer circumference of vessel, holding the guard securely in place.