This item is now discontinued. Please browse similar items
Glas-Col 099C S23 Tissue Homogenizer Pestle, PTFE, 0.966" dia, straight side
Representative image only.
Glas-Col Mfr # 099C S23 – Item # RK-04368-22
Choose pestle material, shape, and sample size
  • Some pestles fit with more than one tube
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Shaft Diameter (inches)0.966
  • Shaft Diameter (mm)24.5
  • DescriptionTissue Homogenizer Pestle, PTFE, 0.966\" dia, straight side
Key Features
  • Some pestles fit with more than one tube
More About this Item
PTFE Pestles are chemically inert, autoclavable, and resilient. They have smooth, nonwettable surfaces. Straight PTFE pestles have a 1-5/8"L PTFE tip mounted on a 8"L x 1/4" dia notched SS shaft and fit both three-jaw keyless chuck and quick-change chuck.