Gilmont GS-1200-A Micrometer Burette-Style Dispenser; 1/Ea
Representative image only.
Gilmont Item # RK-07846-00
Dispense small volumes at constant velocity—ideal for titrations and fluid injections
$581.69CAD / Each
$111.98CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)2
  • Graduation Interval (mL)0.002
  • DescriptionMicrometer Burette-Style Dispenser; 1/Ea
More About this Item
Turn the handle of this dispenser for precise delivery of small volumes. Dispense fluids with an accuracy of ±0.5% of reading or ±1 scale division, whichever is greater. Fluids only come in contact with the borosilicate glass tip, PTFE syringe barrel, and Viton® O-rings. Optional PTFE O-rings are available separately for applications working with harsh chemicals.
Autoclavable