GF Signet 3-8150-1P(PANEL) 3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
Representative image only.
GF Signet Mfr # 3-8150-1P(PANEL) – Item # RK-05629-22
Use in areas where AC power is not readily available
  • Use in areas where wired power is power is not readily available
$1,144.82CAD / Each
$165.02CAD / Each
for an additional
$165.02CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% of reading
  • Max Operating Temperature (° F)131
  • Max Operating Temperature (° C)55
  • Battery Life4 years nominal
  • Display Type4-digit LCD (upper). Lower: 8-digit LCD
  • Description3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
Key Features
  • Use in areas where wired power is power is not readily available
More About this Item
Independently calibrate flow and totalizer readings. Monitor includes both fixed and resettable totalizers. Features a four-digit flow LCD, and eight-digit totalizer LCD, automatic contrast adjustment, and low-battery indication. Monitor includes unit/multiplier decals for field calibration.
Operates on two internal lithium batteries (included). Use monitor only with the Roto-X™ and Metalex™ flow sensors sold separately. Note: not for use with the Roto-X™ Low-Flow sensors.