General Tools UV513A/B Ultraviolet UVA/B Light Meter

Representative image only.
General Tools Mfr # UV513AB – Item # RK-50513-00
Measure damaging UV (ultraviolet) light
  • New wavelength range of 280 to 400 nm
  • 5 year warranty

PRODUCT OPTIONS

$442.02CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±4% ±1 digit (at 23 ± 5°C)
  • Width (in)2
  • Height (InchShort)6
  • Depth (in)1 1/2
  • Width (cm)5.1
  • Height (cm)15.2
  • Depth (cm)3.8
  • Display TypeBacklit LCD
  • Battery1 9V (included)
  • Intensity (mW/cm2)40
  • DescriptionUltraviolet UVA/B Light Meter
  • Warranty5 years
5 Year WarrantyConformite Europeenne

Key Features

  • New wavelength range of 280 to 400 nm
  • 5 year warranty

More About this Item

Conduct UV degradation studies for pharmaceutical, cosmetic, semiconductor, printing, forensic applications and more. The UV light intensity appears on the large backlit display in either ìW or mW/cm² while the small lower display simultaneously shows minimum, maximum, average, and recorded data. Up to 20 data points can be recorded automatically at a variety of intervals.
The UV-A/B model measures long and medium wave UV-A/B for forensics, chromatography, electrophoresis, and dermatology applications. Use the UV-C model for UV light fume hoods, industrial glass, and nondestructive testing.

Please note: These uits do not sterilize equipment.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat