This item is now discontinued. Please browse similar items
General Tools Extendable, Pivot Joint Magnetic Tool with 5-lb. Pick-up Capacity
Representative image only.
General Tools Mfr # 394 – Item # RK-09914-34
Increase retrieval potential with a magnetic reach
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Adjustable Handle Length13\" to 27\"
  • DescriptionExtendable, Pivot Joint Magnetic Tool with 5-lb. Pick-up Capacity
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These magnetic tools feature high power neodymium magnets that are six times more powerful than a standard cobalt magnet. Compact size allows these tools to fit in your pocket. For greater reach, models 09914-34, -36, and –38 feature extendable shafts for those out-of-reach objects.