This item is now discontinued. Please browse similar items
General Tools DT4947SD 4-Channel Thermometer with Sd Card Data Logging
Representative image only.
General Tools Mfr # DT4947SD – Item # RK-37803-07
Easily transfer data using an SD card from your meter to a PC
  • Adjustable datalogging sampling time settable from one second to one hour
  • Differential temperature between two channels
  • Maximum and minimum readings can be recalled or stored for later use
  • Large backlit LCD
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels4
  • Types Of Probes AcceptedThermocouple, RTD
  • Min Temperature (° F)-148
  • Max Temperature (° F)2372
  • Min Temperature (° C)-100
  • Max Temperature (° C)1300
  • Accuracy±0.4% of reading
  • Resolution0.1°
  • OutputSD card
  • Data LoggingYes
  • Display TypeLCD
  • No. of Probe(s) Included4
  • Battery6 AA (not included)
  • Description4-Channel Thermometer with Sd Card Data Logging
  • Warranty1 year
1 Year Warranty
Key Features
  • Adjustable datalogging sampling time settable from one second to one hour
  • Differential temperature between two channels
  • Maximum and minimum readings can be recalled or stored for later use
  • Large backlit LCD
  • 1 year warranty
More About this Item
This 4 channel thermometer records data onto a standard SD memory card, instead of a built-in memory. The advantage of the data being stored on an SD card is when the memory is full the SD card can be simply exchanged out for unlimited data storage. The data is provided in Excel, allowing the user to easily analyze information without the need for additional software. The thermometer has automatic temperature compensation and is designed to work with RTD''s and the six most popular thermocouple types: K, J, T, E, S and R. User settable parameters include; date, time, sampling time, temperature unit of measure, comma or period decimal division, offset and auto power off.