General Tools DM8200 Digtl Manomtr 100PSI 150PSI Max Pressure
Representative image only.
General Tools Mfr # DM8200 – Item # RK-97008-76
Ideal for laboratory research equipment, computer peripherals, pneumatic controls, HVAC and more
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
PRODUCT OPTIONS
$420.28CAD / Each
$223.96 - $331.51CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigtl Manomtr 100PSI 150PSI Max Pressure
  • Warranty1 year
1 Year Warranty
Key Features
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
More About this Item
Displays gauge or differential pressure.