This item is now discontinued. Please browse similar items
General Tools DAF3009 HVAC/R Anemometer-Psychrometer with CFM/CMM and Enthalpy
Representative image only.
General Tools Mfr # DAF3009 – Item # RK-10001-14
Install, test, and fine-tune your HVAC/R system
  • Measure eight environmental conditions including airflow volume and enthalpy—ideal for HVAC technicians
  • Track MIN/MAX/AVE air speed and airflow volume
  • Back-lit LCD is easy to see in any lighting condition
  • Vane cap protects meter when not in use and is conveniently tethered to help prevent getting lost
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHVAC/R Anemometer-Psychrometer with CFM/CMM and Enthalpy
  • Warranty1 year
1 Year Warranty
Key Features
  • Measure eight environmental conditions including airflow volume and enthalpy—ideal for HVAC technicians
  • Track MIN/MAX/AVE air speed and airflow volume
  • Back-lit LCD is easy to see in any lighting condition
  • Vane cap protects meter when not in use and is conveniently tethered to help prevent getting lost
  • 1 year warranty
More About this Item
Measures air speed (in ft/min, m/s, mph, km/hr, or knots), temperature in °F or °C, airflow volume in CFM or CMM, relative humidity, dew point and wet bulb temperatures, wind chill index, and enthalpy in Btu/lb or kJ/kg with this compact meter. See real-time measurements and track MIN/MAX/AVE session values for air speed and airflow volume. Take enthalpy measurements in ductwork to determine the efficiency of HVAC/R systems. This multi-function meter is small enough to fit in a pocket, and the tethered vane cap keeps it safe in a toolbox.