Fluke TL224 Flexible Silicone Test Lead Set
Representative image only.
Fluke Mfr # TL224 – Item # RK-26002-25
High performance at an economical price
  • DMM test leads (red, black) with safety shrouded, standard diameter banana plugs
  • Right angle connector on one end and straight connector on the other
  • Reinforced strain relief
  • 1.5 meter silicone-insulated wire resists heat & cold
  • CAT IV 600 V, CAT III 1000 V, 10 A rating. UL listed
  • One year warranty
PRODUCT OPTIONS
$64.81CAD / Set of 2
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlexible Silicone Test Lead Set
  • Warranty1 Year
1 Year Warranty
Key Features
  • DMM test leads (red, black) with safety shrouded, standard diameter banana plugs
  • Right angle connector on one end and straight connector on the other
  • Reinforced strain relief
  • 1.5 meter silicone-insulated wire resists heat & cold
  • CAT IV 600 V, CAT III 1000 V, 10 A rating. UL listed
  • One year warranty
More About this Item
Various test leads are available to suit your application.
SureGrip™ accessories are designed to improve steadiness in slippery hands. Rubber overmolded surfaces and finger-hugging curves give the user a comfortable, reliable grip on the accessory so they can focus on making an accurate measurement.
Available for sale in the United States and Canada only.