This item is now discontinued. Please browse similar items
Fluke SCC120E Software, cable and carrying case kit
Representative image only.
Fluke Mfr # SCC120E – Item # RK-26086-72
Three test tools in one!
  • Rugged design assures operation in the harshest environments.
  • Model SCC120E - FlukeView Software, cable and carrying case kit
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSoftware, cable and carrying case kit
  • Warranty1 Year
1 Year Warranty
Key Features
  • Rugged design assures operation in the harshest environments.
  • Model SCC120E - FlukeView Software, cable and carrying case kit
  • 1 year warranty
More About this Item
These compact meters feature a dual input digital oscilloscope, two true-rms digital multimeters, and a dual input paperless recorder. Connect-and-view™ triggering provides instant, stable displays—automatically adjusting and correcting signal patterns. Plot, display and store min, max, peak and average values, up to 16 days, while in the recorder mode. Track problem measurements with time and date stamp in recording mode.
Available for sale in the United States and Canada only.