This item is now discontinued. Please browse similar items
Fluke Networks NFC-KIT-CASE 2800145 Networks NFC-KIT-Case Fiber Optic Cleaning Cube Kit with Case
Representative image only.
Fluke Networks Mfr # NFC-KIT-CASE | 2800145 – Item # RK-17791-75
Reduce Troubleshooting
  • Prevent the #1 cause of fiber link failure: end-face contamination
  • Everything needed for precision cleaning of end-faces on both patch cords and inside ports
  • Specially formulated solvent dissolves contaminants, neutralizes static charge and encapsulates
  • particles for removal
  • A range of wipers and swabs enable cleaning of all styles of fiber optic connections
  • By fitting in your pocket, Fiber Optic Cleaning Cards are conveniently perfect for troubleshooting
  • Use the convenient and rugged carrying case to store all of the cleaning components
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNetworks NFC-KIT-Case Fiber Optic Cleaning Cube Kit with Case
Key Features
  • Prevent the #1 cause of fiber link failure: end-face contamination
  • Everything needed for precision cleaning of end-faces on both patch cords and inside ports
  • Specially formulated solvent dissolves contaminants, neutralizes static charge and encapsulates
  • particles for removal
  • A range of wipers and swabs enable cleaning of all styles of fiber optic connections
  • By fitting in your pocket, Fiber Optic Cleaning Cards are conveniently perfect for troubleshooting
  • Use the convenient and rugged carrying case to store all of the cleaning components
More About this Item
Contaminated end-faces are the #1 cause of fiber link failure. Prevent this common problem with Fluke Networks’ Fiber Optic Cleaning Kits and components. These precision fiber optic cleaning kits and components allow effective removal of all contaminants from end-faces on both patch cords and
inside ports.