This item is now discontinued. Please browse similar items
Fluke FLK-V3003 FC Kit Wireless Kit with AC and DC Voltage Modules
Representative image only.
Fluke Mfr # FLK-V3003 FC KIT – Item # RK-20047-93
Economical kits save you time and money!
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote DC voltage measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurement s remotely at your convenience
  • Logging function allows you to record and save up to 65,000 readings for expanded analysis capabilities
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Kit with AC and DC Voltage Modules
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP54 Rated
Key Features
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote DC voltage measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurement s remotely at your convenience
  • Logging function allows you to record and save up to 65,000 readings for expanded analysis capabilities
  • 3 year warranty
More About this Item
These Fluke connect kits allow you to connect wirelessly to over 20 different Fluke products allowing you to share data without leaving the field. Kits combine wireless modules that allow you to monitor and maintain equipment to keep your business running smoothly.
This Fluke V3003 FC wireless digital multimeter kit with AC and DC voltage clamp modules provides you maximum flexibility for taking measurements for all of your application requirements. Jaw opening of 34 mm accommodates large cables and the wireless system allows you to easily set up measurements once and read them as often and when you want.
Available for sale in USA & Canada only
Whats included:
FC 3000 wireless digital multimeter, V3000 FC AC voltage module, V3001 FC DC voltage module,
TL175, TL222 and TL224 test lead sets, AC175, AC220 and AC285 alligator clip sets, PC3000 FC PC adapter with SW3000 software, C3003 Soft carrying case, three compartment, two magnetic hanging straps for hands-free operation.
Available for sale in the United States and Canada only