This item is now discontinued. Please browse similar items
Fluke FLK-TI400 60HZ/FCA Thermal Imager with 3000 Fc DMM and A3001Fc Iflex Module Kit
Representative image only.
Fluke Mfr # FLK-TI400 60HZ/FCA – Item # RK-39750-77
Taking measurements in the field has never been easier
  • View temperature, mechanical, electrical, and vibrational measurements in one place
  • Share measurements with your entire team without leaving the field
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Detector Size320 x 240
  • Min Temperature (° F)-4
  • Max Temperature (° F)2192
  • Min Temperature (° C)-20
  • Max Temperature (° C)1200
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.09 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.05 at 30
  • DescriptionThermal Imager with 3000 Fc DMM and A3001Fc Iflex Module Kit
  • Warranty2 year
Two year warranty
Key Features
  • View temperature, mechanical, electrical, and vibrational measurements in one place
  • Share measurements with your entire team without leaving the field
  • 2 year warranty
More About this Item
Fluke Connect with ShareLive video - Make the best decisions faster than ever before by viewing all temperature, mechanical, electrical and vibration measurements for each equipment asset in one place. Get started saving time and increasing your productivity. Fluke Connect with ShareLive video call is the only wireless measurement system that lets you stay in contact with your entire team without leaving the field. The Fluke Connect mobile app is available for Android and iOS and works with over 20 different Fluke products.
Kit Includes:
Fluke Ti400 Thermal camera with standard infrared lens
• AC power supply and battery pack charger (including mains adapters)
• Two, rugged lithium ion smart battery packs
• USB cable
• HDMI video cable
• SmartView software available via free download
• Rugged, hard carrying case
• Soft transport bag
• Adjustable hand strap
• Warranty registration card
Fluke 3000 FC Wireless Digital Multimeter
• One (1) a3001 FC Module
• One (1) iFlex 10” coil
• One (1) magnet/strap
• TL175 Test Leads (1 pair)
• AC175 Alligator Clips (1 pair)
• C3003 3-compartment soft case
• Information pack
Available for sale in the United States and Canada only