This item is now discontinued. Please browse similar items
Fluke FLK-LENS/4XTELE2 4x Telephoto Lens
Representative image only.
Fluke Mfr # FLK-LENS/4XTELE2 – Item # RK-39749-28
You've already invested in a Fluke tool, now expand its capabilities with Fluke Genuine Accessories
  • 6.0°H x 4.5°V
  • Compatible with TiX560, TiX520, Ti400, Ti300, and Ti200
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description4x TelePHoto Lens (4607058)
  • Warranty1 year
1 Year Warranty
Key Features
  • 6.0°H x 4.5°V
  • Compatible with TiX560, TiX520, Ti400, Ti300, and Ti200
  • 1 year warranty
More About this Item
These accessories increase the functionality of your test tool and increase your safety and proficiency. With Fluke Genuine Accessories, you can broaden the ability to take your Thermal Imager measurements in different environments to meet your application needs.
View your target magnified 4 times more than a standard lens while staying on the ground or a safe distance away. A 4x telephoto lens can help identify a potential issue in your equipment, even when your target is as tiny and distant as a failed splice on a high electrical line or an overheated refractory on a tall flare stack. Avoid the hassle of sending in your camera to calibrate with the lens — with smart lenses you can just attach the lens to any compatible camera and start taking images. This lens includes lens attachment system that holds the lens securely in place, reducing lens wobble. Made of 100 % diamond-turned germanium with an engineered coating, the most efficient available material to transmit energy to the detector.