This item is now discontinued. Please browse similar items
Fluke FLK-LENS/25MAC2 25 Micron Macro IR Smart Lens for TiX560/520
Representative image only.
Fluke Mfr # FLK-LENS/25MAC2 – Item # RK-39749-27
You've already invested in a Fluke tool, now expand its capabilities with Fluke Genuine Accessories
  • 36.1°H x 27.1°V
  • 8 mm (0.3 in) to 14 mm (0.6 in) with optimal at 10 mm (0.4 in)
  • Compatible with TiX560, TiX520
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description25 Micron Macro IR Smart Lens for Tix560 and Tix520
  • Warranty1 year
1 Year Warranty
Key Features
  • 36.1°H x 27.1°V
  • 8 mm (0.3 in) to 14 mm (0.6 in) with optimal at 10 mm (0.4 in)
  • Compatible with TiX560, TiX520
  • 1 year warranty
More About this Item
These accessories increase the functionality of your test tool and increase your safety and proficiency. With Fluke Genuine Accessories, you can broaden the ability to take your Thermal Imager measurements in different environments to meet your application needs.
Get an incredibly detailed image of very small objects so you can gain insight into thermal buildup and dissipation on printed circuit boards, identify a faulty or undersized component, or potentially even pinpoint precise faulty locations on those components. See details as small as 25 microns, which is smaller than the average human hair. Avoid the hassle of sending in your camera to calibrate with the lens — with smart lenses you can just attach the lens to any compatible camera and start taking images. With your lens attached, use the LaserSharp Auto Focus button to set the focus, then position the camera and target within the working distance of the lens (~8 to ~14 mm). Made of 100 % diamond-turned germanium with an engineered coating, the most efficient available material to transmit energy to the detector. Use the tripod mount thread on the TiX560 or TiX520 to attach the camera to a bench-top mounting system to ensure it remains stable and motionless while capturing images.