DescriptionVibration Sensor Expansion Kit; 1 Yr License
Warranty1 year
Includes
Four vibration sensors, FCCM subscription for each sensor, and epoxy.
Key Features
Remotely measure vibrations in dangerous or hard-to-reach areas for up to three years!
Be alerted to changing conditions to prevent machine failure
Compact and lightweight
IP67 rated—rugged enough for industrial environments
Connect up to 20 sensors to one gateway
Includes one year software license
1 year warranty
More About this Item
Make predictive maintenance fast and easy with the Fluke vibration system consisting of a vibration sensor and gateway. Sensor takes continuous overall vibration measurements of acceleration and velocity for a wide range of machines. Long-term vibration monitoring can help detect imbalance, misalignment, looseness, and bearing faults, saving you catastrophic failures and costly downtime. Install gateway within 65 meters from line-of-sight of sensors. Wi-Fi or mobile router/hotspot connectivity and AC power must be available for gateway to work.
Remotely monitor the conditions using the Fluke Connect Condition Monitoring™ (FCCM) software. Set alarms based on user-defined thresholds or Fluke Overall Vibration Severity (FOVS) scale. FCCM software can send alarms to smart devices or computers within seconds of abnormality. View real-time and historical data to take maintenance action.
Connect up to 20 sensors to one gateway. The number of sensors per gateway is determined by two factors: line of sight and distance from sensors. Select from a starter kit or an expansion kit. Starter kit includes a gateway along with four sensors to get you started on monitoring your equipment. Expansion kit includes four sensors. Each kit is available with 1- or 3-year software licenses.