This item is now discontinued. Please browse similar items
Fluke Calibration 5624-20-M PRT/RTD Probe; 0.25" Dia x 20" L, Mini-Banana
Representative image only.
Fluke Calibration Mfr # 5624-20-M – Item # RK-90449-42
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)32
  • Max Temperature (° F)1832
  • Min Temperature (° C)0
  • Max Temperature (° C)1000
  • Temperature Accuracy±0.011°F (±0.006°C)
  • Connector TypeMini Banana Plug
  • Sheath Diameter (in)0.25
  • DescriptionPRT/RTD Probe; 0.25\" Dia x 20\" L, Mini-Banana
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This unit can reach a temperature of 1000degC with long term drift at 0degC of 10mK and accuracy (including short term stability and calibration uncertainty over the full range) of 55 mK. This pRT has short term stability of 5 mK, and an immersion requirement of less than 6'' (153mm) at 700 degC. Each 5624 comes with a NIST traceable, NVLAP accredited fixed point calibration from 0C to 962C.
Each unit includes a NIST Traceable certificate of calibration, carrying case, and user manual.
FEATURES
  • Temperature range of 0degC to 1000degC
  • Accuracy of ± 0.055degC to 962degC
  • Long-term drift of 0.01degC at 0degC after 100 horus at 1000degC)
Available for sale in the United States and Canada only.