This item is now discontinued. Please browse similar items
Fluke Calibration 5623B-6-D (Hart) Freezer Probe, PRT/RTD, 5-Pin Din, -100 to 156C
Representative image only.
Fluke Calibration Mfr # 5623B-6-D – Item # RK-90449-29
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-148
  • Max Temperature (° F)313
  • Min Temperature (° C)-100
  • Max Temperature (° C)156
  • Temperature Accuracy±0.09°F (±0.05°C)
  • Connector Type5-Pin DIN
  • Description(Hart) Freezer Probe, PRT/RTD, 5-Pin Din, -100 to 156C
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5623B Series - Full immersion freezer PRT/RTD probes
: Use this probe when calibrating freezers, freeze dryers and other cold chambers. This unique freezer probe measures with ±0.05°C accuracy, even when its entire handle and cable are immersed in temperatures as cold as –100°C. Other PRTs will break when the entire assembly is exposed to the temperatures that this probe can handle.
Available for sale in the United States and Canada only.