This item is now discontinued. Please browse similar items
Fluke Calibration 3003240 Industrial PRT Probe, 3.2 mm x 12" , B-Wire
Representative image only.
Fluke Calibration Mfr # 5618B-12-B – Item # RK-90450-43
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)932
  • Min Temperature (° C)-200
  • Max Temperature (° C)500
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeStripped Wire
  • Sheath Diameter (in)0.125
  • DescriptionIndustrial PRT Probe, 3.2 mm x 12\" , B-Wire
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5618B Series – Small Diameter Industrial PRT/RTD probes:
For secondary level performance with full ITS-90 calibration, these PRTs are an excellent choice for critical temperature measurements. Featuring a 3.2 mm diameter (1/8 in) sheath, these industrial standards probes have reduced response time without compromising precision. This small diameter 5618A probe works well in many applications where immersion depth is limited.
Available for sale in the United States and Canada only.